Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask
Products
Product Category
flasks (12)
Brand
Sigma-Aldrich (2)
Manufacturer
Capacity (mL)
Closure Type
Glassware Class
Package Size
Sterility
Style
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z326763
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 100 mL, accuracy: 0.1 mL
Expand
View Pricing
Z326720
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 10 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19
Expand
View Pricing
Z326755
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 50 mL, accuracy: 0.08 mL, neck joint: ST/NS 14/23
Expand
View Pricing
Z326704
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 5 mL, accuracy: 0.04 mL
Expand
View Pricing
Z326739
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 20 mL, accuracy: 0.04 mL
Expand
View Pricing
Z326798
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 250 mL, accuracy: 0.15 mL
Expand
View Pricing
Z326828
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 1,000 mL, accuracy: 0.4 mL
Expand
View Pricing
Z326801
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 500 mL, accuracy: 0.25 mL
Expand
View Pricing
Z326747
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 50 mL, accuracy: 0.06 mL, neck joint: ST/NS 12/21
Expand
View Pricing
Z326771
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 200 mL, accuracy: 0.15 mL
Expand
View Pricing
Z326712
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 25 mL, accuracy: 0.06 mL, neck joint: ST/NS 12/21
Expand
View Pricing
Z326836
BRAND
®
BLAUBRAND
®
ETERNA volumetric flask,
volume 2,000 mL, accuracy: 0.6 mL
Expand
View Pricing