Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
AC adapter for charging dock HandyStep
®
electronic
Synonym(s):
BRAND
®
HandyStep
®
electronic charging base
Products
Product Category
adapters (2)
Manufacturer
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
BR705053
BRAND
®
AC adapter for charging dock HandyStep
®
electronic,
AC/DC input 115 V / 240 V AC, Australian plug
Expand
View Pricing
BR705052
BRAND
®
AC adapter for charging dock HandyStep
®
electronic,
AC/DC input 120 V AC, Japan plug
Expand
View Pricing