Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bovine Serum Albumin Fraction V
CAS Number:
9048-46-8
Products
Product Category
albumins (4)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Tissue
Quality Segment
Geographic Origin
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
810533
Bovine Serum Albumin Fraction V,
pack of 100 g, ≥98%, USA origin, moisture (Karl Fischer) 0-5.0%, Serology: suitable, Blood banking: suitable
Expand
View Pricing
81053N
Bovine Serum Albumin Fraction V,
pack of 10 g, ≥98%, USA origin, Serology: suitable, Blood banking: suitable
Expand
View Pricing
810531
Bovine Serum Albumin Fraction V,
pack of 1 kg, ≥98%, USA origin, moisture (Karl Fischer) 0-5.0%, Serology: suitable, Blood banking: suitable
Expand
View Pricing
810535
Bovine Serum Albumin Fraction V,
pack of 5 kg, ≥98%, USA origin, moisture (Karl Fischer) 0-5.0%, Serology: suitable, Blood banking: suitable
Expand
View Pricing