Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boron nitride
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-20 of 49
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF84056867
Boron nitride,
powder, 10 max. part. size (micron), weight 100 g, purity 99.5%
Expand
View Pricing
GF29548794
Boron nitride,
powder, max. particle size 10 micron, weight 200 g, purity 99.5%
Expand
View Pricing
GF50133934
Boron nitride,
powder, max. particle size 10 micron, weight 500 g, purity 99.5%
Expand
View Pricing
GF26570101
Boron nitride,
sheet, thickness 0.20 mm, size 5 × 5 mm, condition hot-pressed
Expand
View Pricing
GF60773968
Boron nitride,
rod, 4.0 mm diameter, length 100 mm, condition hot-pressed
Expand
View Pricing
GF82111463
Boron nitride,
tube, outside diameter 16 mm, length 50 mm, wall thickness 2.5 mm
Expand
View Pricing
GF89090343
Boron nitride,
tube, outside diameter 6.4 mm, length 100 mm, wall thickness 2.35 mm
Expand
View Pricing
GF43305498
Boron nitride,
rod, 9.0 mm diameter, length 100 mm, condition hot-pressed
Expand
View Pricing
GF85898480
Boron nitride,
sheet, thickness 2.0 mm, diameter 40 mm, condition hot-pressed
Expand
View Pricing
GF62593049
Boron nitride,
sheet, thickness 2.0 mm, diameter 50 mm, condition hot-pressed
Expand
View Pricing
GF13076164
Boron nitride,
rod, 25.4 mm diameter, length 25 mm, condition hot-pressed
Expand
View Pricing
GF17213958
Boron nitride,
sheet, thickness 1.0 mm, 13 mm diameter, condition hot-pressed
Expand
View Pricing
GF33689469
Boron nitride,
sheet, thickness 0.5 mm, 30 mm diameter, condition hot-pressed
Expand
View Pricing
GF36874413
Boron nitride,
sheet, thickness 0.5 mm, size 100 × 100 mm, condition hot-pressed
Expand
View Pricing
GF33022785
Boron nitride,
sheet, thickness 1.0 mm, size 50 × 50 mm, condition pyrolitic
Expand
View Pricing
GF56765807
Boron nitride,
rod, 32 mm diameter, length 50 mm, condition hot-pressed
Expand
View Pricing
GF77998381
Boron nitride,
tube, length 50 mm, outside diameter 6.4 mm, wall thickness 2.35 mm
Expand
View Pricing
GF57864189
Boron nitride,
sheet, thickness 1.0 mm, size 25 × 25 mm, condition pyrolitic
Expand
View Pricing
GF29753781
Boron nitride,
rod, 9.0 mm diameter, length 50 mm, condition hot-pressed
Expand
View Pricing
GF75414911
Boron nitride,
tube, outside diameter 4.0 mm, length 100 mm, wall thickness 1.0 mm
Expand
View Pricing
Page 1 of 3
Page 1 of 3