Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boil proof microtubes
Products
Product Category
tubes (18)
Color
Manufacturer
Sterility
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
AXYMCT150C
Boil proof microtubes,
size 1.5 mL, clear, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200C
Boil proof microtubes,
size 2.0 mL, clear, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150X
Boil proof microtubes,
size 1.5 mL, amber, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150A
Boil proof microtubes,
size 1.5 mL, assorted colors, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200Y
Boil proof microtubes,
size 2.0 mL, yellow, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150SP
Boil proof microtubes,
size 1.5 mL, in spectrum pack, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200A
Boil proof microtubes,
size 2.0 mL, assorted colors, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150Y
Boil proof microtubes,
size 1.5 mL, yellow, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200X
Boil proof microtubes,
size 2.0 mL, amber, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150G
Boil proof microtubes,
size 1.5 mL, green, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150R
Boil proof microtubes,
size 1.5 mL, red, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200R
Boil proof microtubes,
size 2.0 mL, red, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150B
Boil proof microtubes,
size 1.5 mL, blue, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150V
Boil proof microtubes,
size 1.5 mL, violet, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200B
Boil proof microtubes,
size 2.0 mL, blue, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200G
Boil proof microtubes,
size 2.0 mL, green, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200O
Boil proof microtubes,
size 2.0 mL, orange, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200V
Boil proof microtubes,
size 2.0 mL, violet, pkg of 10x500tubes/cs
Expand
View Pricing