Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Nicotinamide adenine dinucleotide hydrate
Synonym(s):
β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide
Empirical Formula (Hill Notation):
C
21
H
27
N
7
O
14
P
2
· xH
2
O
CAS Number:
53-84-9
Molecular Weight:
663.43 (anhydrous basis)
EC Number:
200-184-4
Products
Product Category
vitamins (6)
nucleotides (1)
Brand
Sigma-Aldrich (7)
Sustainability
Biological Source
Color
Feature
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Application
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
43410
β-Nicotinamide adenine dinucleotide hydrate,
≥95% (HPLC)
Expand
View Pricing
N7381
β-Nicotinamide adenine dinucleotide hydrate,
Grade AA-1
Expand
View Pricing
N3014
β-Nicotinamide adenine dinucleotide hydrate,
suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Expand
View Pricing
N7004
β-Nicotinamide adenine dinucleotide hydrate,
≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Expand
View Pricing
N1511
β-Nicotinamide adenine dinucleotide hydrate,
≥99%
Expand
View Pricing
N6522
β-Nicotinamide adenine dinucleotide hydrate,
≥98%, BioUltra, from yeast
Expand
View Pricing
N1636
β-Nicotinamide adenine dinucleotide hydrate,
purified by column chromatography, ≥99%
Expand
View Pricing