Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Lactoglobulin from bovine milk
CAS Number:
9045-23-2
EC Number:
232-928-9
Products
Product Category
proteins (4)
Brand
Sigma-Aldrich (3)
Supelco (1)
Biological Source
Manufacturer
Physical Form
Purity
Format
Tissue
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
certified reference material (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
L3908
β-Lactoglobulin from bovine milk,
≥90% (PAGE), lyophilized powder
Expand
View Pricing
L0130
β-Lactoglobulin from bovine milk,
≥90% (PAGE), lyophilized powder
Expand
View Pricing
L-046
β-Lactoglobulin from bovine milk,
certified reference material, ampule of 100 mg, Cerilliant
®
Expand
View Pricing
L2506
β-Lactoglobulin from bovine milk,
≥85% (PAGE), lyophilized powder
Expand
View Pricing