Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Black plastic caps
Products
Product Category
caps (8)
Brand
Sigma-Aldrich (1)
Supelco (7)
Diameter (mm)
Closure Type
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z106437
Black plastic caps,
Solid top, PTFE-faced rubber liner, thread for 15-425, pkg of 200 ea
Expand
View Pricing
Z188808
Black plastic caps,
Solid top, white rubber liner, thread for 8-425, pkg of 1000 ea
Expand
View Pricing
Z106429
Black plastic caps,
Solid top, PTFE-faced rubber liner, thread for 13-425, pkg of 200 ea
Expand
View Pricing
Z106402
Black plastic caps,
Solid top, white rubber liner, thread for 13-425, pkg of 200 ea
Expand
View Pricing
Z106410
Black plastic caps,
Solid top, white rubber liner, thread for 15-425, pkg of 200 ea
Expand
View Pricing
Z164313
Black plastic caps,
Solid top, PTFE-faced rubber liner, thread for 20-400, pkg of 100 ea
Expand
View Pricing
Z188832
Black plastic caps,
Solid top, PTFE-faced rubber liner, thread for 18-400, pkg of 200 ea
Expand
View Pricing
Z203610
Black plastic caps,
Solid-top, PTFE-faced rubber liner, thread for 15-415
Expand
View Pricing