Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bismuth-lead alloy, Bi55.5Pb44.5
Products
Manufacturer
Physical Form
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF56924466
Bismuth-lead alloy, Bi55.5Pb44.5,
100g lump, 100mm max. lump size
Expand
View Pricing
GF36701287
Bismuth-lead alloy, Bi55.5Pb44.5,
200g lump, 100mm max. lump size
Expand
View Pricing
GF55213882
Bismuth-lead alloy, Bi55.5Pb44.5,
50g lump, 100mm max. lump size
Expand
View Pricing
GF05735134
Bismuth-lead alloy, Bi55.5Pb44.5,
1000g lump, 100mm max. lump size
Expand
View Pricing
GF02043919
Bismuth-lead alloy, Bi55.5Pb44.5,
500g lump, 100mm max. lump size
Expand
View Pricing