Skip to Content
Merck
Biphenyl-4-sulfonyl chloride

Biphenyl-4-sulfonyl chloride

Biphenyl-4-sulfonyl chloride
Synonym(s):
(4-Biphenylyl)sulfonyl chloride, 4-(Chlorosulfonyl)biphenyl, 4-Phenylbenzenesulfonic acid chloride, 4-Phenylbenzenesulfonyl chloride, p-Biphenylsulfonyl chloride, p-Phenylbenzenesulfonyl chloride
Linear Formula:
C6H5C6H4SO2Cl
CAS Number:
Molecular Weight:
252.72
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing