Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Biotainer
®
media bottles
Products
Product Category
bottles (12)
Manufacturer
Capacity (mL)
Diameter (mm)
Height (mm)
ID (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z377058
Biotainer
®
media bottles,
capacity 500 mL, clear PETG
Expand
View Pricing
Z377104
Biotainer
®
media bottles,
capacity 4000 mL, translucent high-density polyethylene
Expand
View Pricing
Z377082
Biotainer
®
media bottles,
capacity 2000 mL, clear PETG
Expand
View Pricing
Z377031
Biotainer
®
media bottles,
capacity 125 mL, clear polycarbonate
Expand
View Pricing
Z377074
Biotainer
®
media bottles,
capacity 1000 mL, clear polycarbonate
Expand
View Pricing
TMO3423-42
Biotainer
®
media bottles,
clear polycarbonate, 20000 mL capacity, case of 3 ea
Expand
View Pricing
Z377023
Biotainer
®
media bottles,
capacity 125 mL, clear PETG
Expand
View Pricing
Z377090
Biotainer
®
media bottles,
capacity 2000 mL, clear polycarbonate
Expand
View Pricing
TMO3500-05
Biotainer
®
media bottles,
clear polycarbonate, 5 mL capacity, case of 500 ea
Expand
View Pricing
TMO3110-42
Biotainer
®
media bottles,
PETG, 1000 mL capacity, case of 35 ea internal packs of 5
Expand
View Pricing
TMO3110-35
Biotainer
®
media bottles,
clear PETG, 1000 mL capacity, case of 35 ea bulk pack
Expand
View Pricing
TMO3500-20
Biotainer
®
media bottles,
clear polycarbonate, 20 mL capacity, case of 500 ea
Expand
View Pricing