Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BIOEAZE
™
Ethyl Vinyl Acetate (EVA) Bags
Products
Product Category
bags (4)
Brand
Millipore (4)
Capacity (L)
Volume (ml)
Sterility
Sterilization Method
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1335B
BIOEAZE
™
Ethyl Vinyl Acetate (EVA) Bags,
1 L Ethyl Vinyl Acetate (EVA) sterile bag
Expand
View Pricing
1330B
BIOEAZE
™
Ethyl Vinyl Acetate (EVA) Bags,
volume 20 L, designed for use with Quick Connect Applications, Double Polybagged
Expand
View Pricing
1938B
BIOEAZE
™
Ethyl Vinyl Acetate (EVA) Bags,
volume 5 L, designed for use with Quick Connect Applications
Expand
View Pricing
1521B
BIOEAZE
™
Ethyl Vinyl Acetate (EVA) Bags,
volume 1 L
Expand
View Pricing