Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bio-based Polyether Polyol
Linear Formula:
(C
3
H
6
O)
n
H
2
O
CAS Number:
345260-48-2
Products
Brand
Sigma-Aldrich (4)
Feature
Melting Point (°C)
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
923990
Bio-based Polyether Polyol,
M
n
400-600 Da
Expand
View Pricing
923982
Bio-based Polyether Polyol,
MN = 2600 - 2800 Dalton
Expand
View Pricing
923966
Bio-based Polyether Polyol,
M
n
1800-2200 Da
Expand
View Pricing
923974
Bio-based Polyether Polyol,
M
n
900-1100 Da
Expand
View Pricing