Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bel-Art
®
Write-on
™
Label Tape
Products
Product Category
labels (32)
Manufacturer
Color
Package Size
Filter Products
Showing 1-20 of 32
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
BAF134630600
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, rainbow, width 3/4 in., pk of 6
Expand
View Pricing
BAF134830100
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, white, width 1 in., pk of 3
Expand
View Pricing
BAF134860075
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, green, width 3/4 in., pk of 4
Expand
View Pricing
BAF134800075
Bel-Art
®
Write-on
™
Label Tape,
roll size 15 yard, white, width 3/4 in., pk of 4
Expand
View Pricing
BAF134880050
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, orange, width 1/2 in., pk of 6
Expand
View Pricing
BAF134870075
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, blue, width 3/4 in., pk of 4
Expand
View Pricing
BAF134850075
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, yellow, width 3/4 in., pk of 4
Expand
View Pricing
BAF134880075
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, orange, width 3/4 in., pk of 4
Expand
View Pricing
BAF134830075
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, white, width 3/4 in., pk of 4
Expand
View Pricing
BAF134620300
Bel-Art
®
Write-on
™
Label Tape,
roll size 15 yard, yellow, green, white, width 3/4 in., pk of 3
Expand
View Pricing
BAF134800050
Bel-Art
®
Write-on
™
Label Tape,
roll size 15 yard, white, width 1/2 in., pk of 6
Expand
View Pricing
BAF134860100
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, green, width 1 in., pk of 3
Expand
View Pricing
BAF134840075
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, red, width 3/4 in., pk of 4
Expand
View Pricing
BAF134850050
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, yellow, width 1/2 in., pk of 6
Expand
View Pricing
BAF134830050
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, white, width 1/2 in., pk of 6
Expand
View Pricing
BAF134860050
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, green, width 1/2 in., pk of 6
Expand
View Pricing
BAF134810100
Bel-Art
®
Write-on
™
Label Tape,
roll size 15 yard, yellow, width 1 in., pk of 3
Expand
View Pricing
BAF134800100
Bel-Art
®
Write-on
™
Label Tape,
roll size 15 yard, white, width 1 in., pk of 3
Expand
View Pricing
BAF134840050
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, red, width 1/2 in., pk of 6
Expand
View Pricing
BAF134840100
Bel-Art
®
Write-on
™
Label Tape,
roll size 40 yard, red, width 1 in., pk of 3
Expand
View Pricing
Page 1 of 2
Page 1 of 2