Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BeadBug
™
prefilled tubes, 2.0 mL capacity
Products
Product Category
homogenizers (10)
Manufacturer
Package Size
Sterility
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z763802
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 3.0 mm Zirconium beads, triple-pure, high impact
Expand
View Pricing
Z763799
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 1.5 mm Zirconium beads, triple-pure, high impact
Expand
View Pricing
Z763748
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 0.5 mm Silica glass beads, acid washed
Expand
View Pricing
Z763829
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 2.8 mm stainless steel beads, acid washed
Expand
View Pricing
Z763764
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 0.1 mm Zirconium beads, triple-pure, high impact
Expand
View Pricing
Z763772
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 0.5 mm Zirconium beads, triple-pure, high impact
Expand
View Pricing
Z763721
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 0.1 mm Silica glass beads, acid washed
Expand
View Pricing
Z763756
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 1.0 mm Silica glass beads, acid washed
Expand
View Pricing
Z763780
BeadBug
™
prefilled tubes, 2.0 mL capacity,
with 1.0 mm Zirconium beads, triple-pure, high impact
Expand
View Pricing
Z763810
BeadBug
™
prefilled tubes, 2.0 mL capacity,
Triple-Pure starter kit, with 10 each of 0.1, 0.5, 1.0, 1.5 and 3.0 mm Zirconium beads
Expand
View Pricing