Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Barium sulfate
Synonym(s):
Baryte, Barium sulfate
Linear Formula:
BaSO
4
CAS Number:
7727-43-7
Molecular Weight:
233.39
EC Number:
231-784-4
Products
Product Category
salts (4)
Brand
SAFC (1)
Sigma-Aldrich (3)
Supelco (1)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
243353
Barium sulfate,
ReagentPlus
®
, 99%
Expand
View Pricing
1.01750
Barium sulfate,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, USP
Expand
View Pricing
202762
Barium sulfate,
99.99% trace metals basis
Expand
View Pricing
1.01748
Barium sulfate,
for white standard DIN 5033
Expand
View Pricing
B8675
Barium sulfate,
meets USP testing specifications
Expand
View Pricing