Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bandelin SONOREX
™
SUPER with built-in heating Ultrasonic baths
Products
Product Category
baths (6)
Manufacturer
Capacity (L)
Diameter (mm)
Length (mm)
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z659673
Bandelin SONOREX
™
SUPER with built-in heating Ultrasonic baths,
Bandelin RK 255 H, capacity 5.7 L
Expand
View Pricing
Z659754
Bandelin SONOREX
™
SUPER with built-in heating Ultrasonic baths,
Bandelin RK 510 H, capacity 9.7 L
Expand
View Pricing
Z659800
Bandelin SONOREX
™
SUPER with built-in heating Ultrasonic baths,
Bandelin RK 514 BH, capacity 18.7 L
Expand
View Pricing
Z659584
Bandelin SONOREX
™
SUPER with built-in heating Ultrasonic baths,
Bandelin RK 100 H, capacity 3 L
Expand
View Pricing
Z659517
Bandelin SONOREX
™
SUPER with built-in heating Ultrasonic baths,
Bandelin RK 52H, capacity 1.8 L
Expand
View Pricing
Z659592
Bandelin SONOREX
™
SUPER with built-in heating Ultrasonic baths,
Bandelin RK 103 H, capacity 4 L
Expand
View Pricing