Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Amylase from sweet potato
Synonym(s):
1,4-α-D-Glucan maltohydrolase
CAS Number:
9000-91-3
EC Number:
232-566-1
Products
Brand
Sigma-Aldrich (2)
Sustainability
Biological Source
Feature
Physical Form
Quality Segment
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
A8781
β-Amylase from sweet potato,
Expand
View Pricing
A7005
β-Amylase from sweet potato,
Type I-B, ammonium sulfate suspension, ≥750 units/mg protein (E1%/280)
Expand
View Pricing