Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations
Products
Product Category
cylinders (8)
Manufacturer
Capacity (mL)
Glassware Class
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
DWKCPB0500P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 500 mL
Expand
View Pricing
DWKCPB1000P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 1000 mL , with +/10mL tolerance
Expand
View Pricing
DWKCPB0100P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 100 mL , with +/-1mL tolerance
Expand
View Pricing
DWKCPB0010P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 10 mL , with +/-0.2mL tolerance
Expand
View Pricing
DWKCPB0050P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 50 mL , with +/-0.5mL tolerance
Expand
View Pricing
DWKCPB0250P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 250 mL
Expand
View Pricing
DWKCPB0025P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 25 mL , with +/-0.5mL tolerance
Expand
View Pricing
DWKCPB2000P
Azlon Hexagonal Base Polypropylene Cylinders with Printed Graduations,
class B, capacity 2000 mL , with +/20mL tolerance
Expand
View Pricing