Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ASSET
®
EZ4-ICA Dry Sampler
Synonym(s):
ASSET
®
EZ4 Dry Samplers
Products
Brand
Supelco (2)
Product Line
Application
Package Size
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
28518-U
ASSET
®
EZ4-ICA Dry Sampler,
for sampling isocyanic acid (ICA), for use with for use with LC-MS or LC-MS/MS, pkg of 40 ea, ASSET®
Expand
View Pricing
28517-U
ASSET
®
EZ4-ICA Dry Sampler,
for sampling isocyanic acid (ICA), for use with for use with LC-MS or LC-MS/MS, pkg of 10 ea, ASSET®
Expand
View Pricing