Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column
Products
Brand
Supelco (15)
Separation Technique
ID (mm)
Length (mm)
Quality Segment
Package Size
Pore Size (Å)
Pore size (µm)
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
50639-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
pk of 1 ea, column L × I.D. 150 mm × 1.5 mm
Expand
View Pricing
50652-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
L × I.D. 5 mm × 4.6 mm, pk of 3 ea, guard columns
Expand
View Pricing
50647-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 4.6 mm, pk of 1 ea
Expand
View Pricing
50648-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 150 mm × 4.6 mm, pk of 1 ea
Expand
View Pricing
50644-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 3 mm, pk of 1 ea
Expand
View Pricing
50637-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
pk of 1 ea, column L × I.D. 50 mm × 1.5 mm
Expand
View Pricing
50641-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 2.1 mm, pk of 1 ea
Expand
View Pricing
50642-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 150 mm × 2.1 mm, pk of 1 ea
Expand
View Pricing
50640-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 50 mm × 2.1 mm, pk of 1 ea
Expand
View Pricing
50643-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 50 mm × 3 mm, pk of 1 ea
Expand
View Pricing
50645-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 150 mm × 3 mm, pk of 1 ea
Expand
View Pricing
50638-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 1.5 mm, pk of 1 ea
Expand
View Pricing
50646-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 50 mm × 4.6 mm, pk of 1 ea
Expand
View Pricing
50650-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
L × I.D. 5 mm × 2.1 mm, pk of 3 ea, guard columns
Expand
View Pricing
50651-U
Ascentis
®
Express 90 Å PCS-C18, 2.7 µm HPLC Column,
L × I.D. 5 mm × 3.0 mm, pk of 3 ea, guard columns
Expand
View Pricing