Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Artis Tweezer
Synonym(s):
Artis Tweezer
Products
Product Category
tweezers (8)
Manufacturer
Length (in)
Length (mm)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z742671
Artis Tweezer,
Style AA-SA
Expand
View Pricing
Z742676
Artis Tweezer,
Style 5-SA
Expand
View Pricing
Z742677
Artis Tweezer,
Style 7-SA
Expand
View Pricing
Z742675
Artis Tweezer,
Style 2A-SA
Expand
View Pricing
Z742669
Artis Tweezer,
Style MM-SA
Expand
View Pricing
Z742672
Artis Tweezer,
Style SS-SA
Expand
View Pricing
Z742670
Artis Tweezer,
Style OO-SA
Expand
View Pricing
Z742674
Artis Tweezer,
Style 3C-SA
Expand
View Pricing