Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Anionic Liposomes
Products
Brand
Sigma-Aldrich (28)
Chemical Composition
Quality Segment
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CPS-514
Anionic Liposomes,
Phosphatidylserine (PS)-based -100% DOPS (10mM)
Expand
View Pricing
CPG-514
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - 100% DOPG ( 10mM)
Expand
View Pricing
CPS-505
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (50:50 molar ratio 10mM)
Expand
View Pricing
CPS-507
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (30:70 molar ratio 10mM)
Expand
View Pricing
CPS-502
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (80:20 molar ratio 10mM)
Expand
View Pricing
CPS-501
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (90:10 molar ratio 10mM)
Expand
View Pricing
CPS-508
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (20:80 molar ratio 10mM)
Expand
View Pricing
CPG-507
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (30:70 molar ratio 10mM)
Expand
View Pricing
CPS-512
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (1:99 molar ratio 10mM)
Expand
View Pricing
CPG-508
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (20:80 molar ratio 10mM)
Expand
View Pricing
CPS-503
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (70:30 molar ratio 10mM)
Expand
View Pricing
CPS-509
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (10:90 molar ratio 10mM)
Expand
View Pricing
CPS-513
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (0.5:99.5 molar ratio 10mM)
Expand
View Pricing
CPG-502
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (80:20 molar ratio 10mM)
Expand
View Pricing
CPG-504
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (60:40 molar ratio 10mM)
Expand
View Pricing
CPG-509
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (10:90 molar ratio 10mM)
Expand
View Pricing
CPS-504
Anionic Liposomes,
Phosphatidylserine (PS)-based - DOPS:DOPC (60:40 molar ratio 10mM)
Expand
View Pricing
CPG-503
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (70:30 molar ratio 10mM)
Expand
View Pricing
CPG-506
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (40:60 molar ratio 10mM)
Expand
View Pricing
CPG-511
Anionic Liposomes,
Phosphatidylglycerol (PG)-based - DOPG:DOPC (2:98 molar ratio 10mM)
Expand
View Pricing
Page 1 of 2
Page 1 of 2