Angiotensin Converting Enzyme from rabbit lung
Synonym(s):
ACE, Peptidyl-dipeptidase A, Peptidyldipeptide Hydrolase
CAS Number:
Products
Product Category
Brand
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing