Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Amber storage bottles
Products
Product Category
bottles (8)
Brand
Supelco (8)
Diameter (mm)
Height (mm)
OD (mm)
Volume (ml)
Closure Type
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
23233
Amber storage bottles,
volume 60 mL, O.D. × H 38 mm × 95 mm, thread for 20-400, bottle only, pkg of 12 ea
Expand
View Pricing
23231
Amber storage bottles,
volume 240 mL, O.D. × H 61 mm × 143 mm, thread for 24-400, PTFE/silicone septum, black phenolic hole cap, pkg of 12 ea
Expand
View Pricing
23230-U
Amber storage bottles,
volume 120 mL, O.D. × H 49 mm × 114 mm, thread for 22-400, PTFE/silicone septum, black phenolic hole cap, pkg of 12 ea
Expand
View Pricing
23229
Amber storage bottles,
volume 60 mL, O.D. × H 38 mm × 95 mm, thread for 20-400, PTFE/silicone septum, black phenolic hole cap, pkg of 12 ea
Expand
View Pricing
23232
Amber storage bottles,
volume 30 mL, O.D. × H 32 mm × 80 mm, thread for 20-400, bottle only, pkg of 12 ea
Expand
View Pricing
23234
Amber storage bottles,
volume 120 mL, O.D. × H 49 mm × 114 mm, thread for 22-400, bottle only, pkg of 12 ea
Expand
View Pricing
23235
Amber storage bottles,
volume 240 mL, O.D. × H 61 mm × 143 mm, thread for 24-400, bottle only, pkg of 12 ea
Expand
View Pricing
23228
Amber storage bottles,
volume 30 mL, O.D. × H 32 mm × 80 mm, thread for 20-400, PTFE/silicone septum, black phenolic hole cap, pkg of 12 ea
Expand
View Pricing