
Aluminum phthalocyanine chloride

Synonym(s):
Chloro(29H,31H-phthalocyaninato)aluminum
Empirical Formula (Hill Notation):
C32H16AlClN8
Products
Formula Weight
Melting Point (°C)
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing