Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum/Copper metal matrix composite
Products
Manufacturer
Physical Form
Length (mm)
OD (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF56424304
Aluminum/Copper metal matrix composite,
rod, Al 77.9%/SiC 17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, 15 mm diameter, length 100 mm, condition metal matrix composite
Expand
View Pricing
GF53037637
Aluminum/Copper metal matrix composite,
rod, Al 77.9%/SiC 17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, 15 mm diameter, length 200 mm, condition metal matrix composite
Expand
View Pricing
GF23651399
Aluminum/Copper metal matrix composite,
rod, Al 77.9%/SiC 17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, 15 mm diameter, length 50 mm, condition metal matrix composite
Expand
View Pricing
GF29704631
Aluminum/Copper metal matrix composite,
rod, Al 77.9%/SiC 17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, 15 mm diameter, length 500 mm, condition metal matrix composite
Expand
View Pricing
GF75335519
Aluminum/Copper metal matrix composite,
tube, Al77.9%/SiC17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, outside diameter 25.4 mm, length 100 mm, wall thickness 1.0 mm
Expand
View Pricing
GF16939682
Aluminum/Copper metal matrix composite,
tube, Al 77.9%/SiC 17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, outside diameter 25.4 mm, length 200 mm, wall thickness 1.0 mm
Expand
View Pricing
GF17460716
Aluminum/Copper metal matrix composite,
tube, Al 77.9%/SiC 17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, outside diameter 25.4 mm, length 500 mm, wall thickness 1.0 mm
Expand
View Pricing
GF82589215
Aluminum/Copper metal matrix composite,
tube, Al77.9%/SiC17.8%/Cu 3.3%/Mg 1.2%/Mn 0.4%, outside diameter 25.4 mm, length 1000 mm, wall thickness 1.0 mm
Expand
View Pricing