Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum chloride
Linear Formula:
AlCl
3
CAS Number:
7446-70-0
Molecular Weight:
133.34
EC Number:
231-208-1
Products
Product Category
acids (6)
salts (1)
Brand
Sigma-Aldrich (6)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade
Anhydrous (3)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
237051
Aluminum chloride,
ReagentPlus
®
, 99%
Expand
View Pricing
206911
Aluminum chloride,
reagent grade, 98%
Expand
View Pricing
563919
Aluminum chloride,
anhydrous, powder, 99.999% trace metals basis
Expand
View Pricing
449598
Aluminum chloride,
anhydrous, powder, 99.99% trace metals basis
Expand
View Pricing
294713
Aluminum chloride,
99.99% trace metals basis
Expand
View Pricing
11019
Aluminum chloride,
anhydrous, sublimed, ≥98%
Expand
View Pricing