Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Alumina
Products
Manufacturer
Physical Form
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF29729650
Alumina,
powder, max. particle size 0.1 micron, weight 100 g, condition 99.99% gamma alumina powder.
Expand
View Pricing
GF41348864
Alumina,
powder, max. particle size 0.1 micron, weight 200 g, condition 99.99% gamma alumina powder.
Expand
View Pricing
GF12944190
Alumina,
powder, mean particle size 45 micron, weight 500 g, purity 99.9%
Expand
View Pricing
GF68527187
Alumina,
powder, 0.1 max. part. size (micron), weight 50 g, condition 99.99% gamma alumina powder.
Expand
View Pricing
GF62302531
Alumina,
powder, 0.1 max. part. size (micron), weight 20 g, condition 99.99% gamma alumina powder.
Expand
View Pricing
GF18024511
Alumina,
powder, mean particle size 45 micron, weight 1000 g, purity 99.9%
Expand
View Pricing
GF11664075
Alumina,
sheet, thickness 0.25 mm, size 60 × 60 mm, purity 99%
Expand
View Pricing
GF83466748
Alumina,
sphere, 0.5 mm diameter, 100 pcs, tolerance +/-2.5um
Expand
View Pricing
GF92436026
Alumina,
powder, 45 mean particle size (micron), weight 100 g, purity 99.9%
Expand
View Pricing
GF74339488
Alumina,
powder, 45 mean particle size (micron), weight 50 g, purity 99.9%
Expand
View Pricing
GF61045499
Alumina,
powder, 0.02 mean particle size (micron), weight 100 g, condition nanopowder
Expand
View Pricing
GF90514559
Alumina,
sheet, thickness 2.5 mm, size 50 × 50 mm, purity 99%
Expand
View Pricing
GF54503557
Alumina,
powder, mean particle size 45 micron, weight 200 g, purity 99.9%
Expand
View Pricing
GF16609057
Alumina,
foam, thickness 7 mm, size 50 × 50 mm, porosity 0.86
Expand
View Pricing
GF93095253
Alumina,
sphere, 1.0 mm diameter, 100 pcs, tolerance +/-2.5um
Expand
View Pricing
GF95382771
Alumina,
foam, thickness 12.7 mm, diameter 38 mm, porosity 0.86
Expand
View Pricing
GF06853855
Alumina,
tube, outside diameter 10.0 mm, length 150 mm, wall thickness 1.5 mm
Expand
View Pricing
GF25331903
Alumina,
tube, outside diameter 0.80 mm, length 100 mm, wall thickness 0.3 mm
Expand
View Pricing
GF52769920
Alumina,
sphere, 1.0 mm diameter, 10_pcs, tolerance +/-2.5um
Expand
View Pricing
GF00537207
Alumina,
tube, outside diameter 4.0 mm, length 250 mm, wall thickness 0.75 mm
Expand
View Pricing
Page 1 of 11
Page 1 of 11