Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
three-neck round-bottom flask
Products
Product Category
flasks (28)
Brand
Sigma-Aldrich (28)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z550337
Aldrich
®
three-neck round-bottom flask,
angled, capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
Z418641
Aldrich
®
three-neck round-bottom flask,
angled, capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
Z122459
Aldrich
®
three-neck round-bottom flask,
angled, capacity 100 mL, joint: ST/NS 14/20
Expand
View Pricing
Z516627
Aldrich
®
three-neck round-bottom flask,
angled, capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z421987
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 22,000 mL, center joint: ST/NS 45/50, side Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z516813
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 1,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z418668
Aldrich
®
three-neck round-bottom flask,
angled, capacity 250 mL, joint: ST/NS 14/20
Expand
View Pricing
Z516767
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z516635
Aldrich
®
three-neck round-bottom flask,
angled, capacity 500 mL, joint: ST/NS 29/32
Expand
View Pricing
Z516643
Aldrich
®
three-neck round-bottom flask,
angled, capacity 1,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z547794
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
Z516724
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 100 mL, joint: ST/NS 14/20
Expand
View Pricing
Z516716
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
Z514608
Aldrich
®
three-neck round-bottom flask,
angled, capacity 250 mL, joint: ST/NS 29/42
Expand
View Pricing
Z421960
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 12,000 mL, center joint: ST/NS 34/45, side Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z421979
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 12,000 mL, center joint: ST/NS 45/50, side Joints: ST/NS 24/40 (2)
Expand
View Pricing
Z516740
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 250 mL, joint: ST/NS 14/20
Expand
View Pricing
Z418676
Aldrich
®
three-neck round-bottom flask,
angled, capacity 500 mL, joint: ST/NS 14/20
Expand
View Pricing
Z514616
Aldrich
®
three-neck round-bottom flask,
angled, capacity 500 mL, joint: ST/NS 29/42
Expand
View Pricing
Z514632
Aldrich
®
three-neck round-bottom flask,
vertical, capacity 250 mL, joint: ST/NS 29/42
Expand
View Pricing
Page 1 of 2
Page 1 of 2