Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
single-neck round-bottom flask
Products
Product Category
flasks (27)
Brand
Sigma-Aldrich (18)
Manufacturer
Capacity (mL)
OD (mm)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z100633
Aldrich
®
single-neck round-bottom flask,
capacity 10 mL, joint: ST/NS 14/20
Expand
View Pricing
Z100676
Aldrich
®
single-neck round-bottom flask,
capacity 100 mL, joint: ST/NS 14/20
Expand
View Pricing
Z100641
Aldrich
®
single-neck round-bottom flask,
capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
Z100668
Aldrich
®
single-neck round-bottom flask,
capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
Z514578
Aldrich
®
single-neck round-bottom flask,
capacity 250 mL, joint: ST/NS 29/42
Expand
View Pricing
Z548871
Aldrich
®
single-neck round-bottom flask,
capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z723134
Aldrich
®
single-neck round-bottom flask,
capacity 5 mL, joint: ST/NS 14/20
Expand
View Pricing
Z548898
Aldrich
®
single-neck round-bottom flask,
capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z548928
Aldrich
®
single-neck round-bottom flask,
capacity 1,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z100684
Aldrich
®
single-neck round-bottom flask,
capacity 250 mL, joint: ST/NS 14/20
Expand
View Pricing
Z548863
Aldrich
®
single-neck round-bottom flask,
capacity 50 mL, joint: ST/NS 29/32
Expand
View Pricing
Z548901
Aldrich
®
single-neck round-bottom flask,
capacity 500 mL, joint: ST/NS 29/32
Expand
View Pricing
Z548944
Aldrich
®
single-neck round-bottom flask,
capacity 3,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z548936
Aldrich
®
single-neck round-bottom flask,
capacity 2,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z548952
Aldrich
®
single-neck round-bottom flask,
capacity 5,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z418633
Aldrich
®
single-neck round-bottom flask,
capacity 22,000 mL, joint: ST/NS 55/50
Expand
View Pricing
Z418609
Aldrich
®
single-neck round-bottom flask,
capacity 12,000 mL, joint: ST/NS 45/50
Expand
View Pricing
Z100692
Aldrich
®
single-neck round-bottom flask,
capacity 500 mL, joint: ST/NS 14/20
Expand
View Pricing
Z418625
Aldrich
®
single-neck round-bottom flask,
capacity 22,000 mL, joint: ST/NS 45/50
Expand
View Pricing
Z414484
Aldrich
®
single-neck round-bottom flask,
capacity 50 mL, joint: ST/NS 24/40
Expand
View Pricing
Page 1 of 2
Page 1 of 2