Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Schlaker reaction tube
Products
Product Category
tubes (7)
Brand
Sigma-Aldrich (7)
Binding Type
Capacity (mL)
Color
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z409235
Aldrich
®
Schlaker reaction tube,
capacity 10 mL, plain joint
Expand
View Pricing
Z409251
Aldrich
®
Schlaker reaction tube,
capacity 50 mL, plain joint
Expand
View Pricing
Z409294
Aldrich
®
Schlaker reaction tube,
capacity 10 mL, screw-thread joint
Expand
View Pricing
Z409324
Aldrich
®
Schlaker reaction tube,
capacity 100 mL, screw-thread joint
Expand
View Pricing
Z409278
Aldrich
®
Schlaker reaction tube,
capacity 100 mL, plain joint
Expand
View Pricing
Z409243
Aldrich
®
Schlaker reaction tube,
capacity 25 mL, plain joint
Expand
View Pricing
Z409308
Aldrich
®
Schlaker reaction tube,
capacity 25 mL, screw-thread joint
Expand
View Pricing