Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Liebig condenser
Products
Brand
Sigma-Aldrich (8)
Height (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z122416
Aldrich
®
Liebig condenser,
Overall H 200 mm, joint: ST/NS 14/20
Expand
View Pricing
Z530948
Aldrich
®
Liebig condenser,
Overall H 60 mm, joint: ST/NS 14/20
Expand
View Pricing
Z530964
Aldrich
®
Liebig condenser,
Overall H 200 mm, joint: ST/NS 24/40
Expand
View Pricing
Z530980
Aldrich
®
Liebig condenser,
Overall H 300 mm, joint: ST/NS 14/20
Expand
View Pricing
Z530956
Aldrich
®
Liebig condenser,
Overall H 100 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531014
Aldrich
®
Liebig condenser,
Overall H 400 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531056
Aldrich
®
Liebig condenser,
Overall H 600 mm, joint: ST/NS 24/40
Expand
View Pricing
Z530999
Aldrich
®
Liebig condenser,
Overall H 300 mm, joint: ST/NS 24/40
Expand
View Pricing