Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
large separatory funnel
Products
Product Category
funnels (5)
Brand
Sigma-Aldrich (5)
Capacity (L)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z102628
Aldrich
®
large separatory funnel,
overall capacity 22 L, glass stopcock
Expand
View Pricing
Z127841
Aldrich
®
large separatory funnel,
capacity 4 L, overall capacity 4 L, PTFE stopcock, joint: ST/NS 45/40
Expand
View Pricing
Z102601
Aldrich
®
large separatory funnel,
overall capacity 12 L, glass stopcock
Expand
View Pricing
Z127876
Aldrich
®
large separatory funnel,
overall capacity 10 L, PTFE stopcock, joint: ST/NS 45/40
Expand
View Pricing
Z127868
Aldrich
®
large separatory funnel,
overall capacity 6 L, PTFE stopcock, joint: ST/NS 45/40
Expand
View Pricing