Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
large-capacity cold trap
Products
Brand
Sigma-Aldrich (9)
Capacity (mL)
OD (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z103101
Aldrich
®
large-capacity cold trap,
bulb capacity 500 mL, joint: ST/NS 55/50
Expand
View Pricing
Z284521
Aldrich
®
large-capacity cold trap,
bulb capacity 1,000 mL, with 50 mm O-ring joint
Expand
View Pricing
Z203750
Aldrich
®
large-capacity cold trap,
Inside finger for traps with ST/NS: 55/50 joint
Expand
View Pricing
Z284505
Aldrich
®
large-capacity cold trap,
bulb capacity 1,000 mL, joint: ST/NS 55/50
Expand
View Pricing
Z183954
Aldrich
®
large-capacity cold trap,
Inside finger for traps with 50mm O-ring
Expand
View Pricing
Z284548
Aldrich
®
large-capacity cold trap,
Replacement bulb, for Z284521, with 50 mm O-ring joint
Expand
View Pricing
Z106909
Aldrich
®
large-capacity cold trap,
bulb capacity 500 mL, with 50 mm O-ring joint
Expand
View Pricing
Z118311
Aldrich
®
large-capacity cold trap,
Replacement bulb, for Z103101, joint: ST/NS 55/50
Expand
View Pricing
Z127396
Aldrich
®
large-capacity cold trap,
Replacement bulb, for Z106909, with 50 mm O-ring joint
Expand
View Pricing