Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Essentials test tube with rim
Products
Product Category
tubes (3)
Brand
Sigma-Aldrich (3)
Manufacturer
Diameter (mm)
OD (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z740989
Aldrich
®
Essentials test tube with rim,
capacity 25 mL, O.D. × L 18 mm × 150 mm, round bottom
Expand
View Pricing
Z740988
Aldrich
®
Essentials test tube with rim,
capacity 16 mL, O.D. × L 15 mm × 125 mm, round bottom
Expand
View Pricing
Z740986
Aldrich
®
Essentials test tube with rim,
capacity 10 mL, O.D. × L 15 mm × 125 mm, round bottom
Expand
View Pricing