Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth
Products
Product Category
flasks (10)
Brand
Sigma-Aldrich (10)
Cap
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z740656
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 500 mL, stopper joint: 7, volume 100-500 mL, accuracy: 50 mL
Expand
View Pricing
Z740663
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 5000 mL, stopper joint: 10, volume 1000-5000 mL, accuracy: 500 mL
Expand
View Pricing
Z740662
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 3000 mL, stopper joint: 10, volume 1000-3000 mL, accuracy: 500 mL
Expand
View Pricing
Z740658
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 1000 mL, stopper joint: 9, volume 250-1000 mL, accuracy: 50 mL
Expand
View Pricing
Z740654
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 250 mL, stopper joint: 6, volume 50-200 mL, accuracy: 25 mL
Expand
View Pricing
Z740652
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 100 mL, stopper joint: 5, volume 50-100 mL, accuracy: 25 mL
Expand
View Pricing
Z740648
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 25 mL, stopper joint: 0, volume 10-25 mL, accuracy: 5 mL
Expand
View Pricing
Z740660
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 2000 mL, stopper joint: 10, volume 600-1900 mL, accuracy: 100 mL
Expand
View Pricing
Z740661
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 2000 mL, stopper joint: 10, volume 600-1900 mL, accuracy: 100 mL
Expand
View Pricing
Z740650
Aldrich
®
Essentials Erlenmeyer flask, narrow mouth,
capacity 50 mL, stopper joint: 1, volume 20-50 mL, accuracy: 10 mL
Expand
View Pricing