Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Dean-Stark trap
Products
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z202541
Aldrich
®
Dean-Stark trap,
capacity 12 mL, joint: ST/NS 14/20
Expand
View Pricing
Z423033
Aldrich
®
Dean-Stark trap,
capacity 100 mL, joint: ST/NS 24/40
Expand
View Pricing
Z516260
Aldrich
®
Dean-Stark trap,
capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z516279
Aldrich
®
Dean-Stark trap,
capacity 300 mL, joint: ST/NS 29/32
Expand
View Pricing
Z516252
Aldrich
®
Dean-Stark trap,
capacity 25 mL, joint: ST/NS 29/32
Expand
View Pricing
Z423068
Aldrich
®
Dean-Stark trap,
capacity 500 mL, joint: ST/NS 24/40
Expand
View Pricing
Z423041
Aldrich
®
Dean-Stark trap,
capacity 300 mL, joint: ST/NS 24/40
Expand
View Pricing
Z423025
Aldrich
®
Dean-Stark trap,
capacity 25 mL, joint: ST/NS 24/40
Expand
View Pricing
Z423076
Aldrich
®
Dean-Stark trap,
capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing