Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
calibrated chromatography column with solvent reservoir
Products
Product Category
columns (12)
Brand
Sigma-Aldrich (12)
OD (mm)
Volume (ml)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z560669
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 2000 mL, top joint: threaded 45/50, column volume 2000 mL
Expand
View Pricing
Z560502
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 250 mL, top joint: threaded 24/40, column volume 200 mL
Expand
View Pricing
Z560472
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 100 mL, top joint: threaded 24/40, column volume 100 mL
Expand
View Pricing
Z560634
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 1000 mL, top joint: threaded 45/40, column volume 1000 mL
Expand
View Pricing
Z560480
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 100 mL, top joint: threaded 29/32, column volume 100 mL
Expand
View Pricing
Z560529
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 500 mL, top joint: threaded 24/40, column volume 400 mL
Expand
View Pricing
Z560510
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 250 mL, top joint: threaded 29/32, column volume 200 mL
Expand
View Pricing
Z560537
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 500 mL, top joint: threaded 29/32, column volume 400 mL
Expand
View Pricing
Z560553
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 500 mL, top joint: threaded 24/40, column volume 600 mL
Expand
View Pricing
Z560626
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 500 mL, top joint: threaded 29/32, column volume 600 mL
Expand
View Pricing
Z560650
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 2000 mL, top joint: threaded 45/40, column volume 2000 mL
Expand
View Pricing
Z560642
Aldrich
®
calibrated chromatography column with solvent reservoir,
reservoir volume 1000 mL, top joint: threaded 45/50, column volume 1000 mL
Expand
View Pricing