Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Ketoglutaric acid
Synonym(s):
2-Oxoglutaric acid, 2-Oxopentanedioic acid
Linear Formula:
HOOCCH
2
CH
2
COCOOH
CAS Number:
328-50-7
Molecular Weight:
146.10
EC Number:
206-330-3
Products
Product Category
substrates (2)
building blocks (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Puriss (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
K1128
α-Ketoglutaric acid,
BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
K1750
α-Ketoglutaric acid,
≥98.5% (NaOH, titration)
Expand
View Pricing
75890
α-Ketoglutaric acid,
99.0-101.0% (T)
Expand
View Pricing
61234
α-Ketoglutaric acid,
analytical standard
Expand
View Pricing