![α-Hydroxy-γ-butyrolactone technical grade](/deepweb/assets/sigmaaldrich/product/structures/698/360/879ce50a-84ca-4370-8eee-d1445d76b138/640/879ce50a-84ca-4370-8eee-d1445d76b138.png)
α-Hydroxy-γ-butyrolactone
![α-Hydroxy-γ-butyrolactone technical grade](/deepweb/assets/sigmaaldrich/product/structures/698/360/879ce50a-84ca-4370-8eee-d1445d76b138/640/879ce50a-84ca-4370-8eee-d1445d76b138.png)
Synonym(s):
4,5-Dihydro-3-hydroxy-2(3H)-furanone
Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing