Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acetonitrile solution
Synonym(s):
Acetonitrile solution
Linear Formula:
CH
3
CN
CAS Number:
75-05-8
Molecular Weight:
41.05
Products
Product Category
solvents (5)
Brand
Sigma-Aldrich (1)
Supelco (4)
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.59002
Acetonitrile solution,
suitable for LC/MS, LiChrosolv
®
, contains 0.1% (v/v) formic acid
Expand
View Pricing
4.80672
Acetonitrile solution,
suitable for HPLC, LiChrosolv
®
, contains 0.05% (v/v) trifluoroacetic acid
Expand
View Pricing
4.80448
Acetonitrile solution,
HPLC grade, suitable for HPLC, LiChrosolv
®
, contains 0.1% (v/v) trifluoroacetic acid
Expand
View Pricing
1.59014
Acetonitrile solution,
suitable for LC/MS, LiChrosolv
®
, contains 0.1% (v/v) trifluoroacetic acid
Expand
View Pricing
900686
Acetonitrile solution,
≥99.6%, suitable for mass spectrometry (MS), contains 0.1 % (v/v) formic acid
Expand
View Pricing