Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acetone-d
6
Synonym(s):
Hexadeuteroacetone, Acetone-D6
Linear Formula:
CD
3
COCD
3
CAS Number:
666-52-4
Molecular Weight:
64.12
EC Number:
211-563-9
Products
Product Category
solvents (6)
stable isotopes (3)
Brand
Sigma-Aldrich (7)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Package Size
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
151793
Acetone-d
6
,
99.9 atom % D
Expand
View Pricing
1.00021
Acetone-d
6
,
deuteration degree min. 99.9% for NMR spectroscopy MagniSolv
™
Expand
View Pricing
444863
Acetone-d
6
,
99.9 atom % D
Expand
View Pricing
434531
Acetone-d
6
,
99.9 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing
175862
Acetone-d
6
,
"100%", 99.95 atom % D
Expand
View Pricing
454133
Acetone-d
6
,
99.9 atom % D, contains 1 % (v/v) TMS
Expand
View Pricing
236853
Acetone-d
6
,
99.5 atom % D
Expand
View Pricing