
α-Boswellic acid

Synonym(s):
(3α,4β)-3-Hydroxyolean-12-en-23-oic acid, 3α-Hydroxyolean-12-en-24-oic acid
Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing