Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Amylase from
Aspergillus oryzae
Synonym(s):
1,4-α-D-Glucan-glucanohydrolase
EC Number:
232-588-1
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (4)
Sustainability
Biological Source
Feature
Physical Form
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
10065
α-Amylase from
Aspergillus oryzae
,
powder, ~30 U/mg
Expand
View Pricing
A9857
α-Amylase from
Aspergillus oryzae
,
≥150 units/mg protein (biuret)
Expand
View Pricing
86250
α-Amylase from
Aspergillus oryzae
,
powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Expand
View Pricing
A8220
α-Amylase from
Aspergillus oryzae
,
aqueous solution, ≥800 FAU/g
Expand
View Pricing