
5-Methoxytryptamine

Synonym(s):
2-(5-Methoxyindol-3-yl)ethylamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-MOT, 5-Methoxyindole-3-ethanamine, Deacetylmelatonin, NSC 56422
Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing