Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Bromo-4-chloro-3-indolyl phosphate
p
-toluidine salt
Synonym(s):
BCIP
®
p
-toluidine salt, X-phosphate
p
-toluidine salt
Empirical Formula (Hill Notation):
C
8
H
6
BrClNO
4
P · C
7
H
9
N
CAS Number:
6578-06-9
Molecular Weight:
433.62
EC Number:
229-506-1
Products
Product Category
substrates (4)
bioactive small molecules (1)
Brand
Sigma-Aldrich (4)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
B8503
5-Bromo-4-chloro-3-indolyl phosphate
p
-toluidine salt,
chromogenic, ≥99% (HPLC), powder
Expand
View Pricing
B6777
5-Bromo-4-chloro-3-indolyl phosphate
p
-toluidine salt,
BioReagent, ≥99%, powder, suitable for molecular biology
Expand
View Pricing
B0274
5-Bromo-4-chloro-3-indolyl phosphate
p
-toluidine salt,
tablet
Expand
View Pricing
16670
5-Bromo-4-chloro-3-indolyl phosphate
p
-toluidine salt,
≥95.0% (HPLC), powder
Expand
View Pricing