Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5,5-Dimethyl-1,3-cyclohexanedione
Synonym(s):
Dimedone, Methone
Linear Formula:
(CH
3
)
2
C
6
H
6
(=O)
2
CAS Number:
126-81-8
Molecular Weight:
140.18
EC Number:
204-804-4
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D153303
5,5-Dimethyl-1,3-cyclohexanedione,
95%
Expand
View Pricing
38490
5,5-Dimethyl-1,3-cyclohexanedione,
for HPLC derivatization, for the determination of aldehyde formaldehyde, ≥99.0%
Expand
View Pricing