Skip to Content
Merck
4,5-Diaminopyrimidine 95%

4,5-Diaminopyrimidine

4,5-Diaminopyrimidine 95%
Empirical Formula (Hill Notation):
C4H6N4
CAS Number:
Molecular Weight:
110.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing