
4,4′-Biphenyldiboronic acid

Synonym(s):
4,4′-(Dihydroxyboryl)-1,1′-biphenyl, 4,4′-Biphenylenediboronic acid
Linear Formula:
(HO)2BC6H4C6H4B(OH)2
CAS Number:
Molecular Weight:
241.84
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing